3-Fluorobenzylamine 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: (3-Fluorophenyl)methanamine

PC3680-100G PC3680-25G PC3680-5G
77414-800EA 174.22 USD
77414-800 77414-802 77414-804
3-Fluorobenzylamine 95%
3-Fluorobenzylamine
Formula: C₇H₈FN
MW: 125.15 g/mol
Boiling Pt: 87…89 °C/25 mmHg
Density: 1.097 g/cm³ (25 °C)
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00008113
CAS Number: 100-82-3
EINECS: 202-891-3
UN: 2735
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR