5-Bromo-3-chloro-2-fluorobenzyl bromide 95%

Supplier: APOLLO SCIENTIFIC

PC303454-1G PC303454-250MG
77414-404EA 633.7 USD
77414-404 77414-406
5-Bromo-3-chloro-2-fluorobenzyl bromide 95%
5-Bromo-3-chloro-2-fluorobenzyl bromide
Formula: C₇H₄Br₂ClF
MW: 302.366 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32640076
CAS Number: 2092617-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR