Methyl 2-{[3-chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}benzoate

Supplier: APOLLO SCIENTIFIC

PC300862-1G PC300862-5G
77414-056EA 176.26 USD
77414-056 77414-058
Methyl 2-{[3-chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}benzoate
Methyl 2-{[3-chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}benzoate
Formula: C₁₄H₉ClF₃NO₃
MW: 331.674 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03644664
CAS Number: 852952-21-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR