2-(5-Bromo-2-fluorophenyl)acetonitrile 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC500188-10G PC500188-5G
77415-998EA 95.76 USD
77415-998 77416-000
2-(5-Bromo-2-fluorophenyl)acetonitrile 98%
2-(5-Bromo-2-fluorophenyl)acetonitrile
Formula: C₈H₅BrFN
MW: 214.037 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09804785
CAS Number: 305800-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR