2,2,3,4,4,4-Hexafluoro-1-butanol 95%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
1H,1H,3H-Perfluoro-1-butanol, 2,2,3,4,4,4-Hexafluorobutan-1-ol
PC4653-500G
PC4653-100G
PC4653-25G
77415-654EA
126.16
USD
77415-654
77415-650
77415-652
2,2,3,4,4,4-Hexafluoro-1-butanol 95%
2,2,3,4,4,4-Hexafluoro-1-butanol
Contains up to 5% CF3CF(CH2OH)CHF2
Formula:
C₄H₄F₆O MW: 182.07 g/mol Boiling Pt: 114…115 °C Density: 1.564 g/cm³ (25 °C) Flash Pt: 51 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042309 CAS Number: 382-31-0 EINECS: 206-842-7 UN: 1987 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...