2,2,3,3,4,4,4-Heptafluorobutylamine hydrochloride

Supplier: APOLLO SCIENTIFIC
Danger

PC450255-1G PC450255-25G PC450255-5G
77415-512EA 34.64 USD
77415-512 77415-514 77415-516
2,2,3,3,4,4,4-Heptafluorobutylamine hydrochloride
2,2,3,3,4,4,4-Heptafluorobutylamine hydrochloride
Formula: C₄H₄F₇N·ClH
MW: 235.531 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22124010
CAS Number: 2794-75-4
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR