3,5-Bis(trifluoromethyl)phenylacetonitrile 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-(3,5-Bis(trifluoromethyl)phenyl)acetonitrile

PC3114G-25G PC3114G-5G PC3114G-100G
77414-494EA 56.64 USD
77414-494 77414-496 77414-492
3,5-Bis(trifluoromethyl)phenylacetonitrile 95%
3,5-Bis(trifluoromethyl)phenylacetonitrile
Formula: C₁₀H₅F₆N
MW: 253.15 g/mol
Density: 1.42 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00009904
CAS Number: 85068-32-2
EINECS: 285-294-0
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR