3-Bromo-4-methyl-5-(trifluoromethyl)phenylacetonitrile 97%

Supplier: APOLLO SCIENTIFIC

PC303602-5G PC303602-1G
77414-414EA 991.3 USD
77414-414 77414-412
3-Bromo-4-methyl-5-(trifluoromethyl)phenylacetonitrile 97%
3-Bromo-4-methyl-5-(trifluoromethyl)phenylacetonitrile
Formula: C₁₀H₇BrF₃N
MW: 278.068 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32640175
CAS Number: 2384884-96-0
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR