2-Chloro-6-fluoro-3-methoxyaniline 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC303030-10G PC303030-1G PC303030-5G
77414-338EA 577.66 USD
77414-338 77414-340 77414-342
2-Chloro-6-fluoro-3-methoxyaniline 98%
2-Chloro-6-fluoro-3-methoxyaniline
Formula: C₇H₇ClFNO
MW: 175.59 g/mol
Melting Pt: 37…40 °C
Storage Temperature: Ambient
MDL Number: MFCD09832265
CAS Number: 1017777-58-0
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR