2,4-Difluorobenzoyl chloride 97%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
PC2723-100G
PC2723-250G
PC2723-25G
77413-770EA
83.54
USD
77413-770
77413-772
77413-774
2,4-Difluorobenzoyl chloride 97%
2,4-Difluorobenzoyl chloride
Formula:
C₇H₃ClF₂O MW: 176.55 g/mol Boiling Pt: 96…98 °C/15 mmHg Density: 1.437 g/cm³ (25 °C) Flash Pt: 82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000658 CAS Number: 72482-64-5 EINECS: 276-690-4 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...