3,5-Difluorobenzonitrile 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC2684-10G PC2684-250G PC2684-50G
77413-758EA 21.26 USD
77413-758 77413-760 77413-762
3,5-Difluorobenzonitrile 98%
3,5-Difluorobenzonitrile
Formula: C₇H₃F₂N
MW: 139.1 g/mol
Boiling Pt: 166 °C
Melting Pt: 86…87 °C
Storage Temperature: Ambient
MDL Number: MFCD00010311
CAS Number: 64248-63-1
EINECS: 264-752-3
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR