2,5-Dibromo-α,α,α-trifluorotoluene 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC2195-100G PC2195-25G PC2195-5G
77413-558EA 74.38 USD
77413-558 77413-560 77413-562
2,5-Dibromo-α,α,α-trifluorotoluene 98%
2,5-Dibromo-α,α,α-trifluorotoluene
Formula: C₇H₃Br₂F₃
Melting Pt: 46…48 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00013554
CAS Number: 7657-09-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR