2-Chloro-6-fluorotoluene 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC1910-100G PC1910-500G
77413-410EA 22.42 USD
77413-410 77413-412
2-Chloro-6-fluorotoluene 97%
2-Chloro-6-fluorotoluene
Formula: C₇H₆ClF
Boiling Pt: 154…156 °C
Density: 1.191 g/cm³ (25 °C)
Flash Pt: 46 °C
Storage Temperature: Ambient
MDL Number: MFCD00000570
CAS Number: 443-83-4
EINECS: 207-141-9
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR