3-Bromo-2-fluoroprop-1-ene

Supplier: APOLLO SCIENTIFIC
Warning

PC450149-1G PC450149-250MG PC450149-5G
77415-456EA 268.96 USD
77415-456 77415-458 77415-460
3-Bromo-2-fluoroprop-1-ene
3-Bromo-2-fluoroprop-1-ene
Formula: C₃H₄BrF
MW: 138.966 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20492668
CAS Number: 35386-83-5
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR