4-Fluoro-3-(trifluoromethyl)phenylacetonitrile 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-fluoro-3-(trifluoromethyl)phenylacetonitrile

PC4377M-10G PC4377M-1G PC4377M-5G
77415-378EA 49.32 USD
77415-378 77415-380 77415-382
4-Fluoro-3-(trifluoromethyl)phenylacetonitrile 98%
4-Fluoro-3-(trifluoromethyl)phenylacetonitrile
Formula: C₉H₅F₄N
MW: 203.14 g/mol
Boiling Pt: 93 °C/10 mmHg
Storage Temperature: Ambient
MDL Number: MFCD00061183
CAS Number: 220239-65-6
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR