2-Fluoro-5-nitroaniline 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC3860-100G PC3860-25G PC3860-5G
77414-972EA 52.98 USD
77414-972 77414-974 77414-976
2-Fluoro-5-nitroaniline 98%
2-Fluoro-5-nitroaniline
Formula: C₆H₅FN₂O₂
MW: 156.12 g/mol
Melting Pt: 99…101 °C
Flash Pt: 91 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007652
CAS Number: 369-36-8
EINECS: 206-720-3
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR