2,3,5,6-Tetrafluorobenzonitrile 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC3839-1G PC3839-25G PC3839-5G
77414-964EA 21.34 USD
77414-964 77414-966 77414-968
2,3,5,6-Tetrafluorobenzonitrile 97%
2,3,5,6-Tetrafluorobenzonitrile
Formula: C₇HF₄N
Boiling Pt: 88…90 °C/45 mmHg
Melting Pt: 33…36 °C
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00012286
CAS Number: 5216-17-1
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR