2-Fluoro-4-methylpyridine 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Fluoro-4-picoline

PC3824-100G PC3824-25G
77414-954EA 45.4 USD
77414-954 77414-956
2-Fluoro-4-methylpyridine 97%
2-Fluoro-4-methylpyridine
Formula: C₆H₆FN
Boiling Pt: 155…156 °C
Density: 1.078 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00041224
CAS Number: 461-87-0
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR