5-Fluoro-2-methoxybenzoyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

PC303006-1G PC303006-5G
77414-312EA 93.74 USD
77414-312 77414-314
5-Fluoro-2-methoxybenzoyl chloride
5-Fluoro-2-methoxybenzoyl chloride
Formula: C₈H₆ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD09832309
CAS Number: 704-03-0
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR