4-Fluoro-2-(trifluoromethoxy)benzonitrile 97%

Supplier: APOLLO SCIENTIFIC

PC302989-1G PC302989-250MG PC302989-5G
77414-300EA 151.8 USD
77414-300 77414-302 77414-304
4-Fluoro-2-(trifluoromethoxy)benzonitrile 97%
4-Fluoro-2-(trifluoromethoxy)benzonitrile
Formula: C₈H₃F₄NO
MW: 205.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19687191
CAS Number: 1323966-32-0
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR