2,3-Difluorobenzyl bromide 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: α-Bromo-2,3-difluorotoluene

PC2739-100G PC2739-25G PC2739-5G
77413-812EA 236.36 USD
77413-812 77413-814 77413-816
2,3-Difluorobenzyl bromide 98%
2,3-Difluorobenzyl bromide
Formula: C₇H₅BrF₂
MW: 207.02 g/mol
Boiling Pt: 186
Melting Pt: 8
Density: 1.628 g/cm³ (25 °C)
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00042488
CAS Number: 113211-94-2
UN: 3265
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR