2,6-Difluorobenzylamine 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC2737-100G PC2737-25G PC2737-5G
77413-800EA 162.4 USD
77413-800 77413-802 77413-804
2,6-Difluorobenzylamine 95%
2,6-Difluorobenzylamine
Formula: C₇H₇F₂N
MW: 143.14 g/mol
Boiling Pt: 174…177 °C
Density: 1.197 g/cm³ (25 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00010144
CAS Number: 69385-30-4
EINECS: 273-983-9
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR