3,5-Difluoroaniline 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC2613-100G PC2613-25G PC2613-500G
77413-716EA 68.26 USD
77413-716 77413-718 77413-720
3,5-Difluoroaniline 98%
3,5-Difluoroaniline
Formula: C₆H₅F₂N
Boiling Pt: 80 °C/20 mmHg
Melting Pt: 40…42 °C
Flash Pt: 75 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007763
CAS Number: 372-39-4
EINECS: 206-752-8
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR