(RS)-1-(3,5-Difluorophenyl)ethylamine 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1-(3,5-Difluorophenyl)ethanamine, 3,5-Difluoro-alpha-methylbenzylamine

PC2002-1G PC2002-25G PC2002-5G
77413-442EA 25.4 USD
77413-442 77413-444 77413-446
(RS)-1-(3,5-Difluorophenyl)ethylamine 97%
(RS)-1-(3,5-Difluorophenyl)ethylamine
Formula: C₈H₉F₂N
MW: 157.16 g/mol
Boiling Pt: 175 °C
Storage Temperature: Ambient
MDL Number: MFCD04038286
CAS Number: 321318-29-0
UN: 1760
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR