Tetrabutylammonium fluoride 1.0 M in THF

Supplier: APOLLO SCIENTIFIC
Danger

PC1945-1L PC1945-250ML
77413-434EA 234.32 USD
77413-434 77413-436
Tetrabutylammonium fluoride 1.0 M in THF
Tetrabutylammonium fluoride
Formula: C₁₆H₃₆FN
MW: 261.47
Boiling Pt: 65…67 °C
Density: 0.903 g/cm³ (25 °C)
Flash Pt: –17 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011747
CAS Number: 429-41-4
EINECS: 207-057-2
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR