2-Methoxy-5-(trifluoromethyl)benzyl bromide

Supplier: APOLLO SCIENTIFIC
Danger

PC302694-1G PC302694-25G PC302694-5G
77414-242EA 33.26 USD
77414-242 77414-244 77414-246
2-Methoxy-5-(trifluoromethyl)benzyl bromide
2-Methoxy-5-(trifluoromethyl)benzyl bromide
Formula: C₉H₈BrF₃O
Melting Pt: 34…37 °C
Storage Temperature: Ambient
MDL Number: MFCD06660318
CAS Number: 685126-88-9
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR