4-Ethoxy-3,5-difluorobenzyl bromide 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC302337-1G PC302337-25G PC302337-5G
77414-158EA 110.04 USD
77414-158 77414-160 77414-162
4-Ethoxy-3,5-difluorobenzyl bromide 98%
4-Ethoxy-3,5-difluorobenzyl bromide
Formula: C₉H₉BrF₂O
MW: 251.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09258691
CAS Number: 1017779-38-2
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR