2-Methyl-3-(trifluoromethyl)phenol 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC302262-5G PC302262-1G PC302262-25G
77414-154EA 88 USD
77414-154 77414-150 77414-152
2-Methyl-3-(trifluoromethyl)phenol 97%
2-Methyl-3-(trifluoromethyl)phenol
Formula: C₈H₇F₃O
Melting Pt: 45…48 °C
Storage Temperature: Ambient
MDL Number: MFCD09832280
CAS Number: 934180-47-9
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR