Ethyl 2-(difluoromethyl)-6-methylimidazo[1,2-a]pyridine-3-carboxylate

Supplier: APOLLO SCIENTIFIC

PC301054-1G PC301054-250MG
77414-080EA 495.16 USD
77414-080 77414-082
Ethyl 2-(difluoromethyl)-6-methylimidazo[1,2-a]pyridine-3-carboxylate
Ethyl 2-(difluoromethyl)-6-methylimidazo[1,2-a]pyridine-3-carboxylate
Formula: C₁₂H₁₂F₂N₂O₂
MW: 254.233 g/mol
Storage Temperature: Ambient
CAS Number: 2514953-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR