2,4-Difluorotoluene 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2,4-Difluoro-1-methylbenzene

PC2930-100G PC2930-25G PC2930-5G
77413-982EA 116.14 USD
77413-982 77413-984 77413-986
2,4-Difluorotoluene 98%
2,4-Difluorotoluene
Formula: C₇H₆F₂
MW: 128.12 g/mol
Boiling Pt: 113…117 °C
Density: 1.12 g/cm³ (25 °C)
Flash Pt: 15 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000332
CAS Number: 452-76-6
EINECS: 207-211-9
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR