2-Amino-6-(trifluoromethyl)benzonitrile

Supplier: APOLLO SCIENTIFIC
Warning

PC2853-1G PC2853-250MG PC2853-5G
77413-918EA 30.4 USD
77413-918 77413-920 77413-922
2-Amino-6-(trifluoromethyl)benzonitrile
2-Amino-6-(trifluoromethyl)benzonitrile
Formula: C₈H₅F₃N₂
Melting Pt: 70…73 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08741381
CAS Number: 58458-11-0
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR