7H-Perfluoro-4-methyl-3,6-dioxaoctanesulfonic acid

Supplier: APOLLO SCIENTIFIC

Synonyms: 1,1,2,2-Tetrafluoro-2-[1,1,1,2,3,3-hexafluoro-3-(1,2,2,2-tetrafluoroethoxy)propan-2-yl]oxyethanesulphonic acid

PC28311-1G PC28311-250MG
77413-906EA 111.24 USD
77413-906 77413-908 77413-910
7H-Perfluoro-4-methyl-3,6-dioxaoctanesulfonic acid
7H-Perfluoro-4-methyl-3,6-dioxaoctanesulfonic acid
Formula: C₇H₂F₁₄O₅S
MW: 464.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32011184
CAS Number: 749836-20-2
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR