5-Fluoropyridine-2-carbonitrile 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC2830-100G PC2830-25G PC2830-5G
77413-894EA 536.92 USD
77413-894 77413-896 77413-904
5-Fluoropyridine-2-carbonitrile 97%
5-Fluoropyridine-2-carbonitrile
Formula: C₆H₃FN₂
Melting Pt: 35…41 °C
Flash Pt: 98.9 °C
Storage Temperature: Ambient
MDL Number: MFCD08741371
CAS Number: 327056-62-2
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR