2,6-Dichloro-4-fluorophenol 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC2433-100G PC2433-25G PC2433-5G
77413-634EA 275.08 USD
77413-634 77413-636 77413-638
2,6-Dichloro-4-fluorophenol 98%
2,6-Dichloro-4-fluorophenol
Formula: C₆H₃Cl₂FO
MW: 180.99 g/mol
Boiling Pt: 142 °C/100 mmHg
Melting Pt: 47…51 °C
Flash Pt: 103 °C
Storage Temperature: Ambient
MDL Number: MFCD00010675
CAS Number: 392-71-2
EINECS: 609-626-4
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR