2,2,3,3,3-Pentafluoropropyl trifluoromethanesulfonate 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC1327-25G PC1327-5G
77413-130EA 330.1 USD
77413-130 77413-132
2,2,3,3,3-Pentafluoropropyl trifluoromethanesulfonate 95%
2,2,3,3,3-Pentafluoropropyl trifluoromethanesulfonate
Formula: C₄H₂F₈O₃S
MW: 282.11 g/mol
Boiling Pt: 102…105 °C/740 mmHg
Density: 1.702 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD01862006
CAS Number: 6401-00-9
UN: 2927
ADR: 6.1+8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR