3-Fluoro-1-propanol 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC1281-25G PC1281-5G
77413-122EA 38.72 USD
77413-122 77413-124
3-Fluoro-1-propanol 97%
3-Fluoro-1-propanol
Formula: C₃H₇FO
Boiling Pt: 127.5…128 °C/40 mmHg
Density: 1.039 g/cm³ (25 °C)
Flash Pt: 20 °C
Storage Temperature: Ambient
MDL Number: MFCD00039548
CAS Number: 462-43-1
UN: 1987
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR