5-(Difluoromethoxy)-2-fluorobenzyl bromide

Supplier: APOLLO SCIENTIFIC
Danger

PC302610-1G PC302610-5G
77414-204EA 216.63 USD
77414-204 77414-206
5-(Difluoromethoxy)-2-fluorobenzyl bromide
5-(Difluoromethoxy)-2-fluorobenzyl bromide
Formula: C₈H₆BrF₃O
MW: 255.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09832361
CAS Number: 1017779-36-0
UN: 3265
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR