4-(Difluoromethoxy)-2-fluorobenzyl bromide 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC302561-1G PC302561-5G PC302561-10G
77414-194EA 109.14 USD
77414-194 77414-196 77414-192
4-(Difluoromethoxy)-2-fluorobenzyl bromide 97%
4-(Difluoromethoxy)-2-fluorobenzyl bromide
Formula: C₈H₆BrF₃O
Storage Temperature: Ambient
MDL Number: MFCD09832360
CAS Number: 1017779-33-7
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR