2-Chloro-3,6-difluorobenzonitrile 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC302072-1G PC302072-25G PC302072-5G
77414-116EA 21.32 USD
77414-116 77414-118 77414-120
2-Chloro-3,6-difluorobenzonitrile 98%
2-Chloro-3,6-difluorobenzonitrile
Formula: C₇H₂ClF₂N
MW: 173.55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660175
CAS Number: 886501-33-3
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR