2,6-Difluoroaniline 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC2610-100G PC2610-25G PC2610-500G
77413-710EA 34.64 USD
77413-710 77413-712 77413-714
2,6-Difluoroaniline 98%
2,6-Difluoroaniline
Formula: C₆H₅F₂N
Boiling Pt: 70…71 °C/40 mmHg
Density: 1.199 g/cm³ (25 °C)
Flash Pt: 43 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007655
CAS Number: 5509-65-9
EINECS: 226-853-0
UN: 1992
ADR: 3+6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR