Diethyl fluoromalonate 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC2564-100G PC2564-25G PC2564-5G
77413-702EA 63.16 USD
77413-702 77413-704 77413-706
Diethyl fluoromalonate 97%
Diethyl fluoromalonate
Formula: C₇H₁₁FO₄
Boiling Pt: 121…122 °C/30 mmHg
Density: 1.129 g/cm³ (25 °C)
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00009139
CAS Number: 685-88-1
EINECS: 211-684-7
UN: 1760
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR