2-(Trifluoromethoxy)phenylacetonitrile 96%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
PC2346-1G
PC2346-25G
PC2346-5G
77413-606EA
21.26
USD
77413-606
77413-608
77413-610
2-(Trifluoromethoxy)phenylacetonitrile 96%
2-(Trifluoromethoxy)phenylacetonitrile
Formula:
C₉H₆F₃NO Boiling Pt: 222…223 °C Density: 1.28 g/cm³ (25 °C) Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00236326 CAS Number: 137218-25-8 UN: 3276 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...