3-Bromo-1,1,1-trifluoroacetone 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC1600-100G PC1600-25G
77413-260EA 116.14 USD
77413-260 77413-262
3-Bromo-1,1,1-trifluoroacetone 97%
3-Bromo-1,1,1-trifluoroacetone
Formula: BrH₂CC(O)CF₃
MW: 190.95 g/mol
Boiling Pt: 85…87 °C
Melting Pt: –80 °C
Density: 1.839 g/cm³ (25 °C)
Flash Pt: 5 °C
Storage Temperature: Ambient
MDL Number: MFCD00039237
CAS Number: 431-35-6
EINECS: 207-071-9
UN: 2924
ADR: 3+8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR