2-Amino-4,5-difluorobenzonitrile

Supplier: APOLLO SCIENTIFIC

PC110108-1G PC110108-500MG PC110108-5G
77412-996EA 172.18 USD
77412-996 77412-998 77413-000
2-Amino-4,5-difluorobenzonitrile
2-Amino-4,5-difluorobenzonitrile
Formula: C₇H₄F₂N₂
MW: 154.117 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09835282
CAS Number: 219823-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR