5-Bromo-2-(trifluoromethyl)pyridine 97%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
5-Bromo-2-trifluoromethylpyridine
PC1054-100G
PC1054-25G
PC1054-5G
77412-924EA
253.68
USD
77412-924
77412-926
77412-928
5-Bromo-2-(trifluoromethyl)pyridine 97%
5-Bromo-2-(trifluoromethyl)pyridine
Formula:
C₆H₃BrF₃N MW: 226.00 g/mol Boiling Pt: 68…70 °C/10 mmHg Melting Pt: 39…45 °C Flash Pt: 78 °C Storage Temperature: Ambient |
MDL Number:
MFCD06657686 CAS Number: 436799-32-5 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...