2-Bromo-5-(trifluoromethyl)aniline 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 3-Amino-4-bromobenzotrifluoride, 6-Bromo-α,α,α-trifluoro-m-toluidine, 3-Amino-4-bromo-α,α,α-trifluorotoluene, 3-Amino-4-brom-α,α,α-trifluortoluol

PC1044-25G PC1044-5G
77412-918EA 28.52 USD
77412-918 77412-920
2-Bromo-5-(trifluoromethyl)aniline 97%
2-Bromo-5-(trifluoromethyl)aniline
Formula: C₇H₅BrF₃N
MW: 240.02
Boiling Pt: 81…82 °C/5 mmHg
Density: 1.675 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007637
CAS Number: 454-79-5
EINECS: 207-227-6
UN: 3267
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR