Potassium vinyltrifluoroborate 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC3373-100G PC3373-25G PC3373-5G
77414-702EA 215.98 USD
77414-702 77414-704 77414-706
Potassium vinyltrifluoroborate 97%
Formula: C₂H₃BF₃K
Melting Pt: 226…230 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD02093335
CAS Number: 13682-77-4
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR