Ethyl chlorodifluoroacetate 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC3170-100G PC3170-25G PC3170-5G
77414-542EA 158.94 USD
77414-542 77414-544 77414-552
Ethyl chlorodifluoroacetate 98%
Ethyl chlorodifluoroacetate
Formula: C₄H₅ClF₂O₂
Boiling Pt: 96…97 °C
Density: 1.252 g/cm³ (25 °C)
Flash Pt: 18 °C
Storage Temperature: Ambient
MDL Number: MFCD00013662
CAS Number: 383-62-0
EINECS: 206-850-0
UN: 2924
ADR: 3+8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR