1,2-Epoxy-3-fluoropropane 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 3-Fluoro-1,2-propenoxide

PC3158-5G PC3158-1G PC3158-25G
77414-534EA 241.46 USD
77414-534 77414-530 77414-532
1,2-Epoxy-3-fluoropropane 98%
1,2-Epoxy-3-fluoropropane
Formula: C₃H₅FO
MW: 76.07 g/mol
Boiling Pt: 85…86 °C
Density: 1.067 g/cm³ (25 °C)
Flash Pt: 4 °C
Storage Temperature: Refrigerator
CAS Number: 503-09-3
EINECS: 207-960-1
UN: 2924
ADR: 3+8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR