3,5-Bis(trifluoromethyl)benzoyl chloride 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC3104-100G PC3104-25G PC3104-5G
77414-462EA 87.62 USD
77414-462 77414-464 77414-466
3,5-Bis(trifluoromethyl)benzoyl chloride 98%
3,5-Bis(trifluoromethyl)benzoyl chloride
Formula: C₉H₃ClF₆O
Boiling Pt: 65…67 °C/12 mmHg
Density: 1.526 g/cm³ (25 °C)
Flash Pt: 72 °C
Storage Temperature: Ambient
MDL Number: MFCD00000387
CAS Number: 785-56-8
EINECS: 212-322-0
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR